Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum Chloride Hexahydrate (Crystalline/USP), Fisher Chemical™

Catalog No. A576500 Shop All Fisherbrand Products
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle

Chemical Identifiers

CAS 7784-13-6
Molecular Formula AlCl3H12O6
Molecular Weight (g/mol) 241.42
MDL Number MFCD00149134
InChI Key JGDITNMASUZKPW-UHFFFAOYSA-K
IUPAC Name aluminium(3+) hexahydrate trichloride
SMILES O.O.O.O.O.O.[Al+3].[Cl-].[Cl-].[Cl-]

Specifications

Melting Point 181°C
Color White
pH 2.0
Physical Form Solid
Quantity 500 g
Formula Weight 241.43
Percent Purity 95.0 to 102.0% (USP)
Grade USP
Identification Pass Test (USP)
Packaging Poly Bottle
Density 2.39g/cm³
Chemical Name or Material Aluminum Chloride Hexahydrate
Show More Show Less
DOTInformation DOT Class 8, : Corrosive
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.