Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum chloride hexahydrate, Reagent Grade

Catalog No. p-4824578
Change view
Click to view available options
Quantity:
500 g
2 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1229736 500 g
AA12297A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1229736 Supplier Thermo Scientific Chemicals Supplier No. 01229736
Only null left
Add to Cart

CAS: 7784-13-6 | AlCl3H12O6 | 241.42 g/mol

Pharmaceutical preparations of aluminum chloride hexahydrate are used to treat hyperhidrosis, a condition involving excessive sweating. It is also employed in deodorants, antiperspirant preparations, dyeing fabrics, refining crude oil, textile finishing, manufacturing parchment paper, preserving the wood and disinfecting stables. It also finds use as a reagent in organic synthesis. Aluminum chloride hexahydrate is also useful in water treatment process, medicine and cosmetic products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7784-13-6
Molecular Formula AlCl3H12O6
Molecular Weight (g/mol) 241.42
MDL Number MFCD00149134
InChI Key JGDITNMASUZKPW-UHFFFAOYSA-K
SMILES O.O.O.O.O.O.[Al+3].[Cl-].[Cl-].[Cl-]

Specifications

Melting Point 100°C (decomposition)
Color White
Physical Form Crystals
Quantity 500 g
Solubility Information Highly soluble in water. Soluble in alcohol,ether,glycerol,propylene glycol and acetonitrile.
IUPAC Name aluminum(3+) hexahydrate trichloride
Formula Weight 241.43 (133.34 Anhydrous)
Grade Reagent
Odor Odorless
Density 2.398
Chemical Name or Material Aluminum chloride hexahydrate
Show More Show Less
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation DOT Class: 8, Packaging Group: III
EINECSNumber 231-208-1
RTECSNumber BD0530000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.