Alfa Aesar™ Aluminum chloride, ultra dry, 99.999% (metals basis)

Manufacturer: Alfa Aesar™ 4431322

Quantity 100g

 View more versions of this product

Catalog No. AA4431322

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7446-70-0
    Molecular Formula AlCl3
    Formula Weight 133.34
    MDL Number MFCD00003422
    UN Number UN1726
    Merck Index 14,337
    Chemical Name or Material Aluminum chloride, Ultra dry

    Specifications

    Formula Weight 133.34
    Melting Point 190°C (subl.)
    Packaging Ampule
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Sensitivity Moisture Sensitive
    UN Number UN1726
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314
    Causes severe skin burns and eye damage.
    Storage Note 1
    • Ampouled under argon
    Odor Pungent
    MDL Number MFCD00003422
    Quantity 100g
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: ALUMINUM CHLORIDE, ANHYDROUS
    RTECS Number BD0525000
    TSCA TSCA
    Molecular Formula AlCl3
    pH 2.4
    Physical Form Powder
    CAS 7446-70-0
    Merck Index 14,337
    Density 2.44g/mL
    Chemical Name or Material Aluminum chloride, Ultra dry
    Assay 99.999%, (metals basis)

    GHS H Statement
    H314
    Causes severe skin burns and eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    • Ampouled under argon

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit