Learn More
Aluminum chloride, anhydrous, Reagent Grade
Description
It is used as a catalyst for Friedel-Crafts acylation and alkylation of aromatic compounds. It is one of the most commonly employed Lewis acids for a wide variety of organic transformations. It catalyzes the ene reaction, polymerization, and isomerization reactions. For example, it can be used to synthesis ethyl benzene which is a precursor for producing polystyrene. It can be used for producing dodecylbenzene, a key intermediate for detergents. It is useful in the production of anthroquinone, the presursor for dyestuffs. It is used in the synthesis of bis(arene) metal complexes, through Fischer-Hafner synthesis. Gattermann-Koch reaction employs aluminum chloride for introducing formyl group onto aromatic rings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 190° (sublimation) |
Color | Gray to Yellow |
Physical Form | Lump/Powder |
Quantity | 5 x 1 kg |
Solubility Information | Highly soluble in water. Soluble in alcohol,ether,glycerol,propylene glycol and acetonitrile. |
Formula Weight | 133.34 |
Grade | Reagent |
Odor | Pungent |
Sensitivity | Moisture sensitive |
Density | 2.44 |
Show More |
Safety and Handling
Hazard Category | H314-H335 |
Hazard Statement | Keep container tightly closed and dry. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). |
Precautionary Statement | P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
DOTInformation | DOT Class: 8, Packaging Group: II |
EINECSNumber | 231-208-1 |
RTECSNumber | BD0525000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.