Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum chloride, anhydrous, Reagent Grade

Catalog No. p-4781759
Click to view available options
Quantity:
250 g
1 kg
5 x 1 kg

CAS: 7446-70-0 | AlCl3 | 133.33 g/mol

It is used as a catalyst for Friedel-Crafts acylation and alkylation of aromatic compounds. It is one of the most commonly employed Lewis acids for a wide variety of organic transformations. It catalyzes the ene reaction, polymerization, and isomerization reactions. For example, it can be used to synthesis ethyl benzene which is a precursor for producing polystyrene. It can be used for producing dodecylbenzene, a key intermediate for detergents. It is useful in the production of anthroquinone, the presursor for dyestuffs. It is used in the synthesis of bis(arene) metal complexes, through Fischer-Hafner synthesis. Gattermann-Koch reaction employs aluminum chloride for introducing formyl group onto aromatic rings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7446-70-0
Molecular Formula AlCl3
Molecular Weight (g/mol) 133.33
MDL Number MFCD00003422
InChI Key VSCWAEJMTAWNJL-UHFFFAOYSA-K
Synonym aluminum chloride anhydrous, aluminum iii chloride, aluminiumchloride, aluminumchloride, aluminum chlorid, aluminum cloride, alumimum chloride, aluminum-chloride, trichloroaluminium, chloride aluminum
IUPAC Name aluminum(3+) trichloride
SMILES [Al+3].[Cl-].[Cl-].[Cl-]

Specifications

Melting Point 190° (sublimation)
Color Gray to Yellow
Physical Form Lump/Powder
Quantity 250 g
Solubility Information Highly soluble in water. Soluble in alcohol,ether,glycerol,propylene glycol and acetonitrile.
Formula Weight 133.34
Grade Reagent
Odor Pungent
Sensitivity Moisture sensitive
Density 2.44
Chemical Name or Material Aluminum chloride
Show More Show Less
Hazard Category H314-H335
Hazard Statement Keep container tightly closed and dry. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation DOT Class: 8, Packaging Group: II
EINECSNumber 231-208-1
RTECSNumber BD0525000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.