Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum Cyclohexanebutyrate, 95%, Thermo Scientific™

Catalog No. AC400560050
Click to view available options
Quantity:
5 g

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38598-34-4
Molecular Formula C20H35AlO5
Molecular Weight (g/mol) 382.48
MDL Number MFCD00013771
InChI Key JUGYFOMQOWSWDP-UHFFFAOYSA-K

Specifications

Color White
Elemental Analysis 94% min.
Infrared Spectrum Authentic
Assay Percent Range 95%
Linear Formula (C6H11(CH2)3COO)2AlOH
Quantity 5 g
Formula Weight 382.47
Physical Form Crystalline Powder
Chemical Name or Material Aluminum cyclohexanebutyrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.