Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum di(isopropoxide)acetoacetic ester chelate, Al 9.6% min

Catalog No. p-7080482
Click to view available options
Quantity:
100 g
500 g

CAS: 14782-75-3 | C12H23AlO5

Aluminum di(isopropoxide)acetoacetic ester chelate is used as a gel ink. It finds application in paint to give better drying, luster and hardness, and improve the performance as surface wrinkling. It is also used as a crosslinking agent in epoxy, alkyd and polyester resins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14782-75-3
Molecular Formula C12H23AlO5
MDL Number MFCD00135521

Specifications

Density 1.05 g/mL
Flash Point 50°C (122°F)
Odor Alcohol-like
Quantity 100 g
UN Number UN1993
Sensitivity Moisture sensitive
Formula Weight 274.3
Percent Purity ≥9.6%
Physical Form Liquid
Chemical Name or Material Aluminum di (isopropoxide)acetoacetic ester chelate
Hazard Category H226
Hazard Statement GHS H Statement
H226-H315-H319
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 238-851-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.