Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum flake, APS 11 micron, 99.7% (metals basis)

Catalog No. p-4781760
Click to view available options
Quantity:
250 g
1 kg
5 kg

CAS: 7429-90-5 | Al | 26.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7429-90-5
Molecular Formula Al
Molecular Weight (g/mol) 26.98
MDL Number MFCD00134029
InChI Key XAGFODPZIPBFFR-UHFFFAOYSA-N
Synonym aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
PubChem CID 5359268
ChEBI CHEBI:33629
SMILES [Al]

Specifications

Color Silver to Gray
Appearance Silver Gray flakes
Density 2.7g/cm3
Quantity 250 g
Ignition Point 400°C (752°F)
Odor Odorless
Sensitivity Water reactive
Form Flake
Assay Percent Range 99.7% (Metals basis)
IUPAC Name aluminum
Chemical Name or Material Aluminum
Show More Show Less
Hazard Category Contact with water liberates highly flammable gases. Spontaneously flammable in air.
Hazard Statement Keep container tightly closed and dry.
DOTInformation DOT Class: 4.3, Packaging Group: II
EINECSNumber 231-072-3
RTECSNumber BD0330000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.