Learn More
Aluminum fluoride trihydrate, 97%
Description
Aluminum fluoride trihydrate is used as an additive for the production of aluminum by electrolysis. It is also used to inhibit fermentation and in the preparation of low index films. It serves as a flux ingredient for the removal of magnesium in the refining of aluminum scrap. Further, it is used in the manufacture of aluminum silicates, ceramics and glass. In addition, it acts as a catalyst for organic synthesis and filler in glass industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 250°C (in -3H2O) |
Color | White |
Physical Form | Powder |
Quantity | 2 kg |
Merck Index | 14,339 |
Solubility Information | Slightly soluble in water,acids and alkalis. Insoluble in alcohol and acetone. |
Formula Weight | 138.02 (83.98 Anhydrous) |
Percent Purity | 97% |
Odor | Odorless |
Density | 2.17 g/mL |
Show More |
Safety and Handling
Hazard Category | H315-H320-H335 |
Hazard Statement | GHS H Statement H301-H315-H319-H335 Toxic if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
EINECSNumber | 232-051-1 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.