Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum foil, 1.5mm (0.06 in.) thick, annealed, 99% (metals basis), Si and Fe (combined) typically 1%, Thermo Scientific™

Catalog No. p-7059364
Click to view available options
Quantity:
100 x 100 mm
100 x 500 mm

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7429-90-5
Molecular Formula Al
Molecular Weight (g/mol) 26.98
MDL Number MFCD00134029
InChI Key XAGFODPZIPBFFR-UHFFFAOYSA-N
Synonym aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
PubChem CID 5359268
ChEBI CHEBI:33629
IUPAC Name aluminum
SMILES [Al]

Specifications

Quantity 100 x 100 mm
Odor Odorless
Weight ≈10.125g/50x50mm
Form Foil
Percent Purity 99%
Concentration or Composition (by Analyte or Components) Si and Fe (combined) typically 1%
Assay (metals basis)
Chemical Name or Material Aluminum foil, 1.5mm (0.06 in.) thick, Annealed, Temper: soft
EINECSNumber 231-072-3
TSCA Yes
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.