Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Aluminum Foil Tape

Catalog No. p-7182397
Click to view available options
Dimensions (L x W):
60 yd. x 1 in.
60 yd. x 2 in.
Quantity:
24 Rolls/Cs.
36 Rolls/Cs.

3M™ Aluminum Foil Tape is ideal for numerous applications across a broad range of industries.

  • Thermally conductive tape helps dissipate heat and improves heating and cooling efficiency to protect temperature sensitive materials
  • Resistant to chemicals to protect surfaces during de-paint process and other chemical masking operations
  • Highly engineered acrylic adhesive provides long term durability, but removes cleanly after harsh masking applications
  • Comprised of a dead soft aluminum foil backing and coated with a highly engineered pressure sensitive acrylic adhesive
  • Ideal for heat shielding, heat reflecting, chemical masking, light enhancement, chemical milling, seaming, sealing and de-paint operations.
  • Withstands exposure to flame, weathering, moisture and UV
  • Heat and light reflective tape protects surfaces and enhances lighting efficiency
  • Dead soft aluminum foil backing conforms to curved and uneven surfaces
  • Recognized by UL to UL 746C and Classified by UL to UL 723, Class L for low flammability rating
  • Contains plastic core center
TRUSTED_SUSTAINABILITY

Specifications

Product Type Aluminum Foil Tape
For Use With (Application) Painting, Stripping
Adhesive Type Acrylic
Color Silver
Dimensions (L x W) 60 yd. x 1 in.
Material Acrylic
Model 425
Quantity 36 Rolls/Cs.
Thickness (English) 4.6 mil
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.