Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum hydroxide, extra pure, powder

Catalog No. p-4510112
Click to view available options
Quantity:
25 g
500 g
2 kg
10 kg
Packaging:
Plastic bottle
Plastic drum

CAS: 21645-51-2 | AlH3O3 | 78.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21645-51-2
Molecular Formula AlH3O3
Molecular Weight (g/mol) 78.00
MDL Number MFCD00003420
InChI Key WNROFYMDJYEPJX-UHFFFAOYSA-K
IUPAC Name aluminium(3+) trihydroxide
SMILES [OH-].[OH-].[OH-].[Al+3]

Specifications

Color White
pH 8.5 to 10 (5% aq. suspension)
Physical Form Powder
Quantity 25 g
Linear Formula Al(OH)3
Merck Index 15,338
Solubility Information Solubility in water: insoluble
Formula Weight 78
Percent Purity ≥63.5% (Al2O3)
Grade Extra Pure
Loss on Ignition 36.5% max.
Packaging Plastic bottle
Chemical Name or Material Aluminum hydroxide
Show More Show Less
EINECSNumber 244-492-7

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.