Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aluminum ICP Standard, 10,000 ppm Al in 5% HNO3, Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
50 mL
100 mL
500 mL
Packaging:
Natural LDPE Bottle
Natural Poly Bottle
Specifications
Specifications
| Packaging | Natural LDPE Bottle |
| Concentration or Composition Notes | 10,000ppm Al |
| CAS Min % | 81.0, 11.23, 6.06 |
| CAS Max % | 84.0, 11.69, 6.31 |
| Equivalence | 1 mL = 10 mg Al |
| Physical Form | Liquid |
| Color | Colorless |
| pH | < 2 |
| Type | Standard |
| Quantity | 100 mL |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction