Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum ICP Standard, 10,000 ppm Al in 5% HNO3, Ricca Chemical
SDP

Catalog No. PAL10KN500 Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
50 mL
100 mL
500 mL
Packaging:
Natural LDPE Bottle
Natural Poly Bottle

Chemical Identifiers

CAS 7784-27-2, 7697-37-2, 7732-18-5
Molecular Formula AlH18N3O18
Molecular Weight (g/mol) 375.129
InChI Key SWCIQHXIXUMHKA-UHFFFAOYSA-N
PubChem CID 24567
IUPAC Name aluminum;trinitrate;nonahydrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3]

Specifications

Packaging Natural Poly Bottle
Concentration or Composition Notes 10,000ppm Al
CAS Min % 81.0, 11.23, 6.06
CAS Max % 84.0, 11.69, 6.31
Equivalence 1 mL = 10 mg Al
Physical Form Liquid
Color Colorless
pH < 2
Type Standard
Quantity 500 mL
Chemical Name or Material Aluminum ICP Standard
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.