Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum iodide, 95%

Catalog No. p-4407191
Click to view available options
Quantity:
25 g
100 g

CAS: 7784-23-8 | AlI3 | 407.695 g/mol

Aluminum iodide is employed as a catalyst to break certain kinds of C-O and N-O bonds. It cleaves aryl ethers and deoxygenates epoxides It can also be used for the manufacture of iodinated organic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7784-23-8
Molecular Formula AlI3
Molecular Weight (g/mol) 407.695
MDL Number MFCD00003427
InChI Key CECABOMBVQNBEC-UHFFFAOYSA-K
Synonym aluminum iodide, aluminum triiodide, aluminum iii iodide, aluminum iodide anhydrous, aluminum iodide, technical grade, aluminum triiodide, aluminum iodide metals basis, aluminum iodide, anhydrous, powder trace metals basis
PubChem CID 82222
IUPAC Name triiodoalumane
SMILES [Al](I)(I)I

Specifications

Boiling Point 360°C
Melting Point 191°C
Physical Form Chunks
Quantity 25 g
Assay Percent Range 95%
Solubility Information Soluble in alcohol,ether,carbon disulfide
Formula Weight 407.69
Sensitivity Light and moisture sensitive
Density 3.98
Chemical Name or Material Aluminum iodide
Hazard Category H314-H335
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 232-054-8
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.