Aluminum iodide, 95%, Alfa Aesar™

Manufacturer: Alfa Aesar 3071414

Quantity 25g

 View more versions of this product

Catalog No. AA3071414

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7784-23-8
    Physical Form Granular
    Assay Percent Range 95%
    Molecular Formula AlI3
    Formula Weight 407.69
    MDL Number MFCD00003427
    Chemical Name or Material Aluminum iodide

    Specifications

    Formula Weight 407.69
    Quantity 25g
    EINECS Number 232-054-8
    Physical Form Granular
    Density 3.98
    Assay Percent Range 95%
    Molecular Formula AlI3
    MDL Number MFCD00003427
    CAS 7784-23-8
    Chemical Name or Material Aluminum iodide

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit