Alfa Aesar™ Aluminum iodide, ultra dry, 99.999% (metals basis)

Manufacturer: Alfa Aesar™ 4717006

Quantity 5g

 View more versions of this product

Catalog No. AA4717006

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7784-23-8
    Formula Weight 407.69
    MDL Number MFCD00003427
    UN Number UN3260
    Merck Index 14,346
    Chemical Name or Material Aluminum iodide, Ultra dry

    Specifications

    Formula Weight 407.69
    Packaging Ampule
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Sensitivity Moisture & Light Sensitive
    Merck Index 14,346
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    DOT Information Transport Hazard Class: 8; Packaging Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
    TSCA TSCA
    MDL Number MFCD00003427
    UN Number UN3260
    Boiling Point 360°C
    Color White to brownish-purple
    Density 3.98g/mL
    Odor Pungent
    Physical Form Powder
    CAS 7784-23-8
    Melting Point 191°C
    Quantity 5g
    Health Hazard 1 Danger
    Storage Note 1
    • Ampouled under argon
    Chemical Name or Material Aluminum iodide, Ultra dry
    Assay 99.999%, (metals basis)

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    • Ampouled under argon

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit