Alfa Aesar™ Aluminum isopropoxide, 99.99+% (metals basis)

Manufacturer: Alfa Aesar™ 2298222

Quantity 100g

 View more versions of this product

Catalog No. AA2298222

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 555-31-7
    Formula Weight 204.25
    MDL Number MFCD00008870
    UN Number UN3181
    Merck Index 14,347
    Chemical Name or Material Aluminum isopropoxide

    Specifications

    Formula Weight 204.25
    Health Hazard 3 GHS P Statement
    P210-P280-P240-P241-P370+P378a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    Use explosion-proof electrical/ventilating/lighting/ equipment.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Health Hazard 2 GHS H Statement
    H228
    Flammable solid.
    RTECS Number BD0975000
    Sensitivity Moisture Sensitive
    MDL Number MFCD00008870
    UN Number UN3181
    Boiling Point 135°C (10mmHg)
    Physical Form Powder
    Quantity 100g
    Merck Index 14,347
    Density 1.035g/mL
    Melting Point 131° to 135°C
    TSCA TSCA
    CAS 555-31-7
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.
    Refractive Index 1.0346
    Chemical Name or Material Aluminum isopropoxide
    Assay ≥99.99%, (metals basis)

    GHS H Statement
    H228
    Flammable solid.

    GHS P Statement
    P210-P280-P240-P241-P370+P378a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    Use explosion-proof electrical/ventilating/lighting/ equipment.
    In case of fire: Use for extinction: CO2, powder or water spray.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit