Learn More
Aluminum metaphosphate, Al{2}O{3} 19.4%, P{2}O{5} 78.9%
Description
Aluminum metaphosphate is used as a constituent of glazes, enamels, glasses and as a high-temperature insulating cement. It is also used as molecular sieves.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | White |
Physical Form | Powder |
Quantity | 1 kg |
Solubility Information | Insoluble in water. |
Formula Weight | 263.9 |
Concentration or Composition (by Analyte or Components) | Al2O3 19.4%, P2O5 78.9% |
Density | 2.779 g/mL |
Chemical Name or Material | Aluminum metaphosphate |
Safety and Handling
Hazard Category | H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.