Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Aluminum Monoclonal Antibody (6F5)
GREENER_CHOICE

Catalog No. PIMA549739
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549739 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549739 Supplier Invitrogen™ Supplier No. MA549739
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Antibody tested in indirect ELISA.

Aluminum interactions with biological systems, particularly plants, can involve gene expression modulation as part of the plant's defense mechanisms. Alterations in gene expression may occur in response to aluminum exposure, which can lead to phytotoxicity and genotoxicity. In plants, mechanisms for aluminum tolerance involve both internal sequestration and external defenses, such as the exudation of organic acids and the regulation of membrane transporters. There are different genetic factors and molecular pathways involved in managing aluminum toxicity, including those influenced by microRNAs and specific gene regulations. The study of these genes and pathways provides insights into plant resilience against environmental stresses caused by metals like aluminum.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Aluminum
Applications ELISA, Immunohistochemistry, Lateral Flow, SDS-Page
Classification Monoclonal
Clone 6F5
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Alias Al
Host Species Mouse
Immunogen Conjugate of Al hapten and KLH.
Purification Method Protein G
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.