Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum n-butoxide, 90+%

Catalog No. AA8775418
Click to view available options
Quantity:
10 g
50 g

CAS: 3085-30-1 | C12H30AlO3 | 249.351 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3085-30-1
Molecular Formula C12H30AlO3
Molecular Weight (g/mol) 249.351
MDL Number MFCD00015237
InChI Key YGVBJFIUURLJDC-UHFFFAOYSA-N
Synonym aluminum iii n-butoxide, aluminum; butan-1-ol
PubChem CID 50933522
IUPAC Name aluminum;butan-1-ol
SMILES CCCCO.CCCCO.CCCCO.[Al]

Specifications

Melting Point 102°C
Physical Form Lump
Quantity 50 g
Linear Formula Al[O(CH2)3CH3]3
UN Number UN3181
Formula Weight 246.32
Percent Purity ≥90%
Sensitivity Moisture sensitive
Chemical Name or Material Aluminum n-butoxide
Hazard Category H228-H315-H319-H335-H336
Hazard Statement GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.
EINECSNumber 221-394-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.