Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum Nitrate Nonahydrate, ≥98%, ACS Reagent, Solstice

Catalog No. 60045463 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-045-463 100 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-045-463 Supplier Solstice Supplier No. 237973100G

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

ACS Reagent,≥98%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7784-27-2
Molecular Formula AlH18N3O18
Molecular Weight (g/mol) 375.129
MDL Number MFCD00149132
InChI Key SWCIQHXIXUMHKA-UHFFFAOYSA-N
Synonym aluminum nitrate nonahydrate, aluminum trinitrate nonahydrate, unii-8mc6621v1h, aluminum iii nitrate, nonahydrate 1:3:9, nitric acid aluminum salt, nitric acid, aluminum salt, nonahydrate 8ci,9ci, aluminum nitrate, acs, aluminum 3+ nonahydrate trinitronate, aluminum nitrate nonahydrate, low mercury, puratronic
PubChem CID 24567
IUPAC Name aluminum;trinitrate;nonahydrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3]

Specifications

Melting Point 73°C
Color White
pH 2.5 to 3.5 at 20°C
Quantity 100 g
Linear Formula Al(NO3)3 · 9H2O
UN Number NONH for all modes of transport
Formula Weight 375.13g/mol
Percent Purity ≥98%
Grade ACS Reagent
Packaging Poly Bottle
Appearance Crystals
Chemical Name or Material Aluminum Nitrate Nonahydrate
Show More Show Less
Hazard Statement P280-P305 + P351 + P338 + P310
Precautionary Statement H318
EINECSNumber 236-751-8
RTECSNumber BD1050000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.