Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium perchlorate, 99+%, for analysis

Catalog No. AC221360010
Click to view available options
Quantity:
25 g
500 g
1 kg
2.5 kg
Packaging:
Plastic bottle

CAS: 7778-74-7 | ClKO4 | 138.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7778-74-7
Molecular Formula ClKO4
Molecular Weight (g/mol) 138.54
MDL Number MFCD00011362
InChI Key YLMGFJXSLBMXHK-UHFFFAOYSA-M
Synonym potassium perchlorate, perchloracap, astrumal, peroidin, perchloric acid, potassium salt, irenat, potassium hyperchloride, perchloric acid, potassium salt 1:1, potassium perchlorate kclo4, potassium perchlorate usp
PubChem CID 516900
IUPAC Name potassium;perchlorate
SMILES [K+].[O-][Cl](=O)(=O)=O

Specifications

Melting Point 400.0°C
Color Colorless to Yellow
Physical Form Liquid
Quantity 1 kg
Assay Percent Range 99+%
Linear Formula KClO4
Merck Index 15,7774
Solubility Information Solubility in water: 17g/L (20°C). Other solubilities: 0.75g/100mL water (0°C),21.8g/100mL water (100°C),practically insoluble in alcohol
Formula Weight 138.55
Percent Purity ≥99%
Grade Analytical
Packaging Plastic bottle
Chemical Name or Material Potassium perchlorate, p.a.
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
May cause fire or explosion; strong oxidizer.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep/Store away from clothing/other combustible materials.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 231-912-9
RTECSNumber SC9700000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.