Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum oxide, 20% in H{2}O, colloidal dispersion

Catalog No. p-4824585
Click to view available options
Quantity:
1 kg
2 x 2.5 kg

Effective in high temperature refractories as a stable binder | CAS: 1344-28-1 | Al2O3 | 101.96 g/mol

Effective in high temperature refractories as a stable binderAluminum oxide, 20% colloidal dispersion is used in dessicators, ceramics, coating materials and electrical insulators. It is an ingredient in sunscreen, and sometimes present in cosmetics such as blush, lipstick, and nail polish. Aluminum oxide is a catalyst in the claus process in hydro desulfurization and some Ziegler-Natta polymerizations. It is widely used as an abrasive, in the automotive industries, cosmetic industries and fiber materials. It is a medium for chromatography in some sodium vapor lamps. It is used in ultrafine acid side polishing slurries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
SMILES [O--].[O--].[O--].[Al+3].[Al+3]

Specifications

Physical Form Particles
Quantity 1 kg
Solubility Information Miscible with ethanol.
IUPAC Name dialuminum;oxygen(2-)
Chemical Name or Material Aluminum oxide
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.