Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum oxide, Aerosol Refractory Paint, Thermo Scientific Chemicals

Catalog No. AA403910CL
Click to view available options
Quantity:
13 oz.
5 x 13 oz.

Provides easy application of uniform, thin, protective high temperature Aluminum oxide coatings. Substrates can be ceramic, graphite or metal. Aluminum oxide is also widely used as an abrasive. In the paper-making process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Provides easy application of uniform, thin, protective high temperature Al2O3 coatings
  • Substrates can be ceramic, graphite or metal
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
IUPAC Name dialuminium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Al+3].[Al+3]

Specifications

Boiling Point -44°C
Color White
Quantity 5 x 13 oz.
UN Number UN1950
Flash Point -104°C (-155°F)
Solubility Information Slightly soluble in water. insoluble in diethyl ether Practically insoluble in ethanol
Packaging Aerosol Spray Can
Chemical Name or Material Aluminum oxide, Aerosol Refractory Paint
Hazard Category H222-H319-H336-H350-X
Hazard Statement GHS H Statement
H222-H319-H336
Extremely flammable aerosol.
Causes serious eye irritation.
May cause drowsiness or dizziness.
Precautionary Statement P201-P202-P261-P264b-P271-P280i-P281-P304+P340-P305+P351+P338-P308+P313-P501c
DOTInformation Transport Hazard Class: 2.1; Packaging Group: ; Proper Shipping Name: AEROSOLS
TSCA Yes
Recommended Storage Ambient temperatures

For Manufacturing and Laboratory Use. Not for beverage use

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.