Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum oxide, gamma-phase, 99.997% (metals basis)

Catalog No. p-7052328
Click to view available options
Quantity:
25 g
100 g
500 g

CAS: 1344-28-1 | Al2O3 | 101.96 g/mol

It is essential in protein extraction as a component in the preparation of tissue. Aluminum oxide is a source of aluminum in reactions, an abrasive agent, and as a refractory material. Furthermore, Aluminum oxide has a relatively high thermal conductivity of 30 Wm-1K-1. Also Aluminum oxide can act as a catalyst in a variety of reactions such as the Claus process and in the dehydration of alcohols to alkenes. It is an inorganic compound used as a grinding or blending agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
IUPAC Name dialuminium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Al+3].[Al+3]

Specifications

Physical Form Powder
Quantity 25 g
Merck Index 14,356
Solubility Information Insoluble in water.
Formula Weight 101.96
Percent Purity 99.997%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Aluminum oxide, gamma-phase
EINECSNumber 215-691-6
RTECSNumber BD1200000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.