Learn More
Aluminum oxide, gamma-phase, 99.997% (metals basis)
Description
It is essential in protein extraction as a component in the preparation of tissue. Aluminum oxide is a source of aluminum in reactions, an abrasive agent, and as a refractory material. Furthermore, Aluminum oxide has a relatively high thermal conductivity of 30 Wm-1K-1. Also Aluminum oxide can act as a catalyst in a variety of reactions such as the Claus process and in the dehydration of alcohols to alkenes. It is an inorganic compound used as a grinding or blending agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Physical Form | Powder |
| Quantity | 25 g |
| Merck Index | 14,356 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 101.96 |
| Percent Purity | 99.997% |
| Odor | Odorless |
| Assay | (metals basis) |
| Chemical Name or Material | Aluminum oxide, gamma-phase |
Safety and Handling
| EINECSNumber | 215-691-6 |
| RTECSNumber | BD1200000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.