Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum oxide, NanoArc™ AL-2220, 30% in mineral spirits, colloidal dispersion with dispersant, Thermo Scientific™

Catalog No. AA4558636
Click to view available options
Quantity:
100 g
500 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Aluminum oxide dispersed in mineral spirits suitable for incorporation in non-polar organic based coatings and formulations where high transparency is important
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
IUPAC Name dialuminium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Al+3].[Al+3]

Specifications

Physical Form Dry Powder
Quantity 500 g
UN Number UN1268
Formula Weight 101.96
Concentration or Composition (by Analyte or Components) 30% in mineral spirits
Chemical Name or Material Aluminum oxide, NanoArc™ AL-2220, Colloidal dispersion with dispersant
Hazard Category Danger
Hazard Statement GHS H Statement
H304-H226
May be fatal if swallowed and enters airways.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
P210-P280-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: PETROLEUM DISTILLATES, N.O.S.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.