Alfa Aesar™ Aluminum oxide, NanoArc™ AL-2220, 30% in mineral spirits, colloidal dispersion with dispersant

Manufacturer: Alfa Aesar™ 4558622

Quantity 100g

 View more versions of this product

Catalog No. AA4558622

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1344-28-1
    Molecular Formula Al2O3
    Formula Weight 101.96g/mol
    MDL Number MFCD00003424
    UN Number UN1268
    Chemical Name or Material Aluminum oxide, NanoArc™ AL-2220, Colloidal dispersion with dispersant

    Specifications

    Concentration 30% in mineral spirits
    Particle Size 20nm APS
    Quantity 100g
    CAS 1344-28-1
    Physical Form Dry powder (typical)
    Molecular Formula Al2O3
    Formula Weight 101.96g/mol
    MDL Number MFCD00003424
    UN Number UN1268
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H304-H226
    May be fatal if swallowed and enters airways.
    Flammable liquid and vapor.
    Health Hazard 3 GHS P Statement
    P210-P280-P301+P310-P303+P361+P353-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: PETROLEUM DISTILLATES, N.O.S.
    Chemical Name or Material Aluminum oxide, NanoArc™ AL-2220, Colloidal dispersion with dispersant

    Description

    • Aluminum oxide dispersed in mineral spirits suitable for incorporation in non-polar organic based coatings and formulations where high transparency is important

    GHS H Statement
    H304-H226
    May be fatal if swallowed and enters airways.
    Flammable liquid and vapor.

    GHS P Statement
    P210-P280-P301+P310-P303+P361+P353-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit