Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum Oxide, Neutral, Brockmann Activity Grade I, Powder, BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Poly Bottle
1344-28-1
Al2O3
101.96
PNEYBMLMFCGWSK-UHFFFAOYSA-N
aluminium oxide,aluminum oxide,alpha-alumina,fasertonerde,abramant,abramax,abrarex,abrasit,aloxite,alundum
9989226
dialuminum;oxygen(2-)
[O-2].[O-2].[O-2].[Al+3].[Al+3]
1344-28-1
Al2O3
101.96
PNEYBMLMFCGWSK-UHFFFAOYSA-N
aluminium oxide,aluminum oxide,alpha-alumina,fasertonerde,abramant,abramax,abrarex,abrasit,aloxite,alundum
9989226
dialuminum;oxygen(2-)
[O-2].[O-2].[O-2].[Al+3].[Al+3]

High quality chemicals for laboratory and specialized industrial use

    For Chromatography
Formula Weight 101.96
Physical Form Powder
Grade BAKER ANALYZED™ Reagent
Packaging Poly Bottle
Specific Gravity 1L = 4.00kg
Quantity 2.5kg
Chemical Name or Material Aluminum Oxide, Neutral, Brockmann Activity Grade I