Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aluminum Oxide, Powder, Chromatography, BAKER ANALYZED™ Reagent, J.T. Baker™
Shop All Avantor Products
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Description
- Suitable for Chromatographic Use
Specifications
Specifications
| Physical Form | Powder |
| Quantity | 500 g |
| Formula Weight | 101.96 |
| Grade | BAKER ANALYZED™ Reagent |
| Packaging | Poly Bottle |
| Specific Gravity | 1L = 4.00kg |
| Chemical Name or Material | Aluminum Oxide |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction