Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum oxide sputtering target, 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick, 99.99% (metals basis), Thermo Scientific™

Catalog No. AA41045KS
Click to view available options
Quantity:
1 Ea.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00003424

Specifications

Quantity 1 Ea.
Solubility Information Insoluble in water.
Formula Weight 101.96
Percent Purity 99.99%
Assay (metals basis)
Chemical Name or Material Aluminum oxide sputtering target, 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick
EINECSNumber 215-691-6
TSCA Yes
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.