Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum phosphate, 97%

Catalog No. AA11109A1
Click to view available options
Quantity:
250 g
1 kg

CAS: 7784-30-7 | AlO4P | 121.951 g/mol

As a flux for ceramics, preparation of dental cement, cement with calcium sulfate and sodium silicate, special glasses, as gel in pharmacy.Aluminum phosphate is used as catalysts, molecular sieves, flux for ceramics and as a gel in pharmacy. It is employed in the preparation of dental cement, emollients and fire-resistant coating. It plays an important role as an anti-fouling agent in textile industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7784-30-7
Molecular Formula AlO4P
Molecular Weight (g/mol) 121.951
MDL Number MFCD00003430
InChI Key ILRRQNADMUWWFW-UHFFFAOYSA-K
Synonym aluminum phosphate, aluminum monophosphate, phosphalugel, phosphaljel, monoaluminum phosphate, aluminum acid phosphate, aluphos, aluminophosphoric acid, aluminiumphosphat german, aluminum phosphate 1:1
PubChem CID 64655
IUPAC Name aluminum;phosphate
SMILES [O-]P(=O)([O-])[O-].[Al+3]

Specifications

Melting Point >1500°C
Color White
Physical Form Powder
Quantity 1 kg
Assay Percent Range 97%
Solubility Information Insoluble in water and acetic acid. Slightly soluble in concentrated hydrochloric acid and nitric acid.Insoluble in water.
Formula Weight 121.95
Appearance White powder
Density 2.566
Chemical Name or Material Aluminum phosphate
Hazard Category H315-H320
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P315-P362-P501c
DOTInformation DOT Class: 8, Packaging Group: III
EINECSNumber 232-056-9
RTECSNumber TB6450000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.