Learn More
Aluminum, plasma standard solution, Specpure™ Al 10,000μg/mL
Description
Aluminum, plasma standard solution is used as a single-element standard solution for plasma emission spectrometry. It is also used as a standard solution in analytical chemistry and atomic absorption spectroscopy.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Physical Form | Liquid |
Concentration | Al in 5% HCl |
UN Number | UN1789 |
Solubility | Miscible with water |
Quantity | 500 mL |
Grade | Specpure™ |
Chemical Name or Material | Aluminum plasma standard solution, Al 10000μg/mL |
Safety and Handling
Hazard Category | H290 |
Hazard Statement | GHS H Statement H314-H335 Causes severe skin burns and eye damage. May cause respiratory irritation. |
Precautionary Statement | P234-P390 |
DOTInformation | Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: HYDROCHLORIC ACID |
EINECSNumber | 231-595-7 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.