Learn More
DWK Life Sciences Kimble™ Aluminum Seal Manual Decrimper
Click to view available options
For Use With (Equipment):
Size 11 Aluminum Vial Seals
Size 13 Aluminum Vial Seals
Size 20 Aluminum Vial Seals
Description
- Manual decrimper for cap size 13 mm
- Crimper part number is 69904-13
Specifications
Specifications
No. per Pack | 1 Each |
Type | Manual Decrimper |
For Use With (Equipment) | Size 13 Aluminum Vial Seals |