Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum, silver coated powder, -200 mesh, 99.9% (metals basis), Ag 19-21 wt%

Catalog No. AA4034014
Click to view available options
Quantity:
25 g
100 g

Catalog No. AA4034014

Supplier: Thermo Scientific Chemicals 04034014

Only null left
Add to Cart

Conductive adhesives | CAS: 11145-27-0 | AlSi | 55.067 g/mol

Conductive adhesives

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 11145-27-0
Molecular Formula AlSi
Molecular Weight (g/mol) 55.067
MDL Number MFCD05865335
InChI Key CSDREXVUYHZDNP-UHFFFAOYSA-N
Synonym silicon aluminum, aluminum-silicon alloy, al.si, aluminum silicon slug, aluminum silicon, aluminum-silane 1/1, aluminum silicon powder,-325 mesh, aluminum silicon slug, 6.35mm 0.25in dia x 6.35mm 0.25in length, aluminum silicon slug, 12.7mm 0.5in dia x 12.7mm 0.5in length, puratronic, aluminum silicon slug, 6.35mm 0.25in dia x 12.7mm 0.5in length, puratronic
PubChem CID 9793685
IUPAC Name aluminum;silicon
SMILES [Al].[Si]

Specifications

Quantity 25 g
Mesh Size −200 Mesh
Odor Odorless
Weight 1.1g/cm3 to 1.7g/cm3
Form Powder
Percent Purity 99.9%
Concentration or Composition (by Analyte or Components) Ag 19 to 21 wt%
Assay (metals basis)
Chemical Name or Material Aluminum, Silver coated powder
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.