Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum Standard, 3750 ppb 3750 μg/L (3.75 ppm), Reagents

Catalog No. 01335895
Click to view available options
Quantity:
4 L

Aluminum Standard, 3750 ppb 3750 μg/L (3.75 ppm), Reagent, use for Aluminum analysis. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Aluminum Standard, 3750 ppb 3750 μg/L (3.75 ppm)
Packaging Poly Bottle
CAS 7732-18-5
Specific Gravity 1.0001
Physical Form Liquid
Color Colorless
pH <7
Boiling Point 100°C
Solubility Completely miscible
Type Conductivity Standard
Industry Type Aluminum Analysis
Quantity 4 L
Grade Reagent
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.