Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aluminum Standard Solution (200 ppm Al) R, Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Natural Poly Bottle
Specifications
Specifications
| Packaging | Natural Poly Bottle |
| Concentration or Composition Notes | 200ppm Al |
| CAS Min % | 97.0, 0.87, 0.31 |
| CAS Max % | 100.0, 0.91, 0.32 |
| Physical Form | Liquid |
| Color | Colorless |
| pH | < 3 |
| Boiling Point | 100°C |
| Quantity | 120 mL |
| Chemical Name or Material | Aluminum Standard Solution, Reagent |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction