Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum Sulfate, 20% w/w, Reagents

Catalog No. 01335894
Change view
Click to view available options
:
Quantity: 4 L
Catalog No.
01-335-894 Quantity: 4 L
1 options

Catalog No. 01-335-894

Supplier: Reagents Holdings Llc CA2075004A

Due to product restrictions, please to purchase or view availability for this product.

Aluminum Sulfate, 20% w/w, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 7784-31-8
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
IUPAC Name water
SMILES O

Specifications

Boiling Point 101°C
Color Colorless
Physical Form Liquid
Quantity 4 L
Percent Purity 20%
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.0893
Chemical Name or Material Aluminum Sulfate, 20% w/w
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.