Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aluminum Sulfate, Crystal, FCC, 99.5-114%, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Click to view available options
Quantity:
45 kg
Packaging:
Fiber Drum
Description
Spectrum™ Chemical Aluminum Sulfate, Crystal, FCC is soluble in water and is mainly used as a thickener and emulsifier in food preparation. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
CAS | 7784-31-8 |
CAS Max % | 100% |
Quantity | 45 kg |
Molecular Formula | Al2H18O21S3 |
UN Number | UN3077 |
InChI Key | KFCWRPFNNIEZRY-UHFFFAOYSA-H |
SMILES | O.O.O.O.O.O.O.O.O.[Al+3].[Al+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O |
IUPAC Name | dialuminium(3+) nonahydrate trisulfate |
Molecular Weight (g/mol) | 504.27 |
Percent Purity | 99.5 to 114% |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction