Learn More
Aluminum sulfate hydrate, 97+%
Description
Aluminum sulfate hydrate is a chemical compound commonly used as a flocculating agent in the purification of drinking water and waste water treatment plants, and also in paper manufacturing. Used as coagulant in decolorization and COD removal of secondary yeast wastewater effluents. It is also plays a significant role in arsenic removal by coagulation process.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | White |
Physical Form | Crystalline |
Quantity | 1 kg |
Assay Percent Range | >97% |
Solubility Information | It is slightly soluble in alcohol and dilute mineral acids. |
Formula Weight | 342.15 (Anhydrous) |
Odor | Odorless |
Appearance | White crystalline |
Density | 1.61 |
Chemical Name or Material | Aluminum sulfate hydrate, hydrate |
Safety and Handling
Hazard Category | H318 |
Precautionary Statement | P280-P305+P351+P338-P310 |
EINECSNumber | 233-135-0 |
RTECSNumber | BD1700000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.