Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum sulfate hydrate, 97+%

Catalog No. p-3256868
Click to view available options
Quantity:
500 g
1 kg
5 kg

CAS: 17927-65-0 | Al2O12S3 | 342.13 g/mol

Aluminum sulfate hydrate is a chemical compound commonly used as a flocculating agent in the purification of drinking water and waste water treatment plants, and also in paper manufacturing. Used as coagulant in decolorization and COD removal of secondary yeast wastewater effluents. It is also plays a significant role in arsenic removal by coagulation process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17927-65-0
Molecular Formula Al2O12S3
Molecular Weight (g/mol) 342.13
MDL Number MFCD00149136
InChI Key DIZPMCHEQGEION-UHFFFAOYSA-H
Synonym aluminum sulfate hydrate, dialuminum trisulfate hydrate, aluminum sulfate usp, aluminum sulfate 2:3 hydrate, alminum sulfate hydrate, aluminum sulfate usan, sulfuric acid, aluminum salt 3:2 , hydrate, 2al.3so4.h2o
PubChem CID 3080712
IUPAC Name dialuminium(3+) trisulfate
SMILES [Al+3].[Al+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Color White
Physical Form Crystalline
Quantity 500 g
Assay Percent Range >97%
Solubility Information It is slightly soluble in alcohol and dilute mineral acids.
Formula Weight 342.15 (Anhydrous)
Odor Odorless
Appearance White crystalline
Density 1.61 g/cm3
Chemical Name or Material Aluminum sulfate hydrate, hydrate
Hazard Category H318
Precautionary Statement P280-P305+P351+P338-P310
EINECSNumber 233-135-0
RTECSNumber BD1700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.