Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum sulfide, 99+%, (metals basis)

Catalog No. AA1285106
Click to view available options
Quantity:
5 g
25 g

Catalog No. AA1285106

Supplier: Thermo Scientific Chemicals 01285106

Only null left
Add to Cart

CAS: 1302-81-4 | Al2S3 | 150.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1302-81-4
Molecular Formula Al2S3
Molecular Weight (g/mol) 150.14
MDL Number MFCD00014162
InChI Key COOGPNLGKIHLSK-UHFFFAOYSA-N
IUPAC Name dialuminium(3+) trisulfanediide
SMILES [Al+3].[Al+3].[S--].[S--].[S--]

Specifications

Melting Point 1100°C
Color Yellow
Physical Form Pieces
Quantity 5 g
Assay Percent Range >99% (Metal basis)
Formula Weight 150.16
Odor Like rotten eggs
Sensitivity Moisture sensitive
Appearance Yellowish granular
Density 2.02 g/cm3
Chemical Name or Material Aluminum sulfide
Show More Show Less
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation DOT Class: 4.3, Packaging Group: III
EINECSNumber 215-109-0
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.