Alfa Aesar™ Aluminum tert-butoxide, 97%

Manufacturer: Alfa Aesar™ 8989114

Quantity 25g

 View more versions of this product

Catalog No. AA8989114

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 556-91-2
    Molecular Formula Al[OC(CH3)3]3
    Formula Weight 246.32
    MDL Number MFCD00008801
    UN Number UN2925
    Beilstein 4148229
    Merck Index 14,333
    Synonym Aluminum tri-tert-butoxide; Tri-tert-butoxyaluminum
    Chemical Name or Material Aluminum tert-butoxide

    Specifications

    Formula Weight 246.32
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Boiling Point 156°C (2mmHg)
    UN Number UN2925
    Sensitivity Moisture Sensitive
    Beilstein 4148229
    Solubility Information Very soluble in organic solvents
    Density 1.0251g/mL
    Odor Alcohol-like
    Physical Form Powder
    Molecular Formula Al[OC(CH3)3]3
    Quantity 25g
    Health Hazard 2 GHS H Statement
    H314-H318-H228
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flammable solid.
    DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, CORROSIVE, ORGANIC, N.O.S.
    TSCA TSCA
    MDL Number MFCD00008801
    CAS 556-91-2
    Merck Index 14,333
    Synonym Aluminum tri-tert-butoxide; Tri-tert-butoxyaluminum
    Health Hazard 1 Danger
    Melting Point 200° to 202°C
    Chemical Name or Material Aluminum tert-butoxide
    Assay 97%

    GHS H Statement
    H314-H318-H228
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flammable solid.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit