Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum tert-butoxide, 97%

Catalog No. p-7080532
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 556-91-2 | C12H27AlO3-3 | 246.327 g/mol

Aluminum tert-butoxide is utilized as a reagent in the Oppenauer oxidation of alcohols to ketones. It is also used in Meerwein-Ponndorf-Verley reduction of ketones. It is actively involved in the preparation of 1,1,2-triethoxyethene by reacting with 1,1,2,2-tetraethoxyethane followed by the elimination of alcohol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 556-91-2
Molecular Formula C12H27AlO3-3
Molecular Weight (g/mol) 246.327
MDL Number MFCD00008801
InChI Key BXQSSDQHLWSBEX-UHFFFAOYSA-N
Synonym aluminum tert-butoxide, acmc-20alth, 2-propanol, 2-methyl-,aluminum salt 3:1
PubChem CID 57350944
IUPAC Name aluminum;2-methylpropan-2-olate
SMILES CC(C)(C)[O-].CC(C)(C)[O-].CC(C)(C)[O-].[Al]

Specifications

Melting Point 200°C to 202°C
Density 1.0251 g/mL
Boiling Point 156°C (2 mmHg)
Odor Alcohol-like
Quantity 5 g
UN Number UN2925
Beilstein 4148229
Sensitivity Moisture sensitive
Merck Index 14,333
Solubility Information Very soluble in organic solvents like alcohols,benzene,toluene and xylene.
Formula Weight 246.32
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Aluminum tert-butoxide
Show More Show Less
Hazard Category H228-H314-H335
Hazard Statement GHS H Statement
H314-H318-H228
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable solid.
Precautionary Statement P210-P240-P241-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, CORROSIVE, ORGANIC, N.O.S.
EINECSNumber 209-146-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.