Learn More
Aluminum tert-butoxide, 97%, Thermo Scientific Chemicals
Description
Aluminum tert-butoxide is utilized as a reagent in the Oppenauer oxidation of alcohols to ketones. It is also used in Meerwein-Ponndorf-Verley reduction of ketones. It is actively involved in the preparation of 1,1,2-triethoxyethene by reacting with 1,1,2,2-tetraethoxyethane followed by the elimination of alcohol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 200°C to 202°C |
Density | 1.0251 g/mL |
Boiling Point | 156°C (2 mmHg) |
Odor | Alcohol-like |
Quantity | 5 g |
UN Number | UN2925 |
Beilstein | 4148229 |
Sensitivity | Moisture sensitive |
Merck Index | 14,333 |
Solubility Information | Very soluble in organic solvents like alcohols,benzene,toluene and xylene. |
Show More |
Safety and Handling
Hazard Category | H228-H314-H335 |
Hazard Statement | GHS H Statement H314-H318-H228 Causes severe skin burns and eye damage. Causes serious eye damage. Flammable solid. |
Precautionary Statement | P210-P240-P241-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, CORROSIVE, ORGANIC, N.O.S. |
EINECSNumber | 209-146-1 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.