Learn More
United Scientific™ Aluminum Tray Dissecting with Wax Layer
SDPSCI_ED
Catalog No. S00348 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Dimensions (L x W):
8 x 6 in.
12 x 8 in.
12 x 10 in.
14 x 10 in.
18 x 12 in.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
S00348 14 x 10 in.
S00344 8 x 6 in.
S00346 18 x 12 in.
S00350 12 x 10 in.
S00352 12 x 8 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. S00348 Supplier United Scientific™ Supplier No. UNDISTRYWX04
Add to Cart
Add to Cart
This item is not returnable. View return policy
Aluminum Tray Dissecting offering an innovative way of securing the sample.
  • Dissecting pans are made of durable aluminum with rounded corners
  • The corners include holes that can be used as tie-down points to hold specimens securely during dissection
  • Trays include a layer of wax
Specifications
Autoclavable Not Autoclavable
Color Silver Tray, Black Wax
Material Aluminum
Dimensions (L x W) 14 x 10 in.