Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ ALX 5407 hydrochloride
GREENER_CHOICESDP

Catalog No. 175710 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-571-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 17-571-0 Supplier Tocris Bioscience™ Supplier No. 1757/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective non-transportable GlyT1 inhibitor

Selective non-transportable inhibitor of the glycine transporter GlyT1 (IC50 values are 3 nM and > 100 μM for human GlyT1c and GlyT2 respectively). Does not recognize other glycine sites, including the glycine site on the NMDA receptor (IC50 > 100 μM).

Racemate also available.

Chemical Identifiers

CAS 200006-08-2
Molecular Formula C24H25ClFNO3

Specifications

Quantity 10 mg
Formula Weight 429.92
Purity >98%
Molecular Weight (g/mol) 429.916
Chemical Name or Material ALX 5407 hydrochloride
Recommended Storage Desiccate at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.