Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ALX1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553876
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553876 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553876 Supplier Invitrogen™ Supplier No. PA553876
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QIQQAKSHFA ATYDISVLPR TDSYPQIQNN LWAGNASGGS VVTSCMLPRD TSSCMTPYSH SPRTDSSYTG FSNHQNQFSH VPLNNFFTDS LLTGATNGHA FETKPEFERR SSSIAVL Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 97%.

Transcriptional activator that acts at a palindromic recognition sequence to enhance the activity of the SV40 and TK promoters. Functions as a repressor with the prolactin promoter in vivo. May play a role in chondrocyte differentiation and may also influence cervix development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALX1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ALX1
Gene Accession No. Q15699
Gene Alias AI314867; ALX homeobox 1; ALX homeobox protein 1; ALX1; C130005I02; Cart1; CART-1; CART1X; cartilage homeo protein 1; cartilage homeoprotein 1; cartilage paired-class homeoprotein 1; epididymis luminal protein 23; FND3; HEL23
Gene Symbols ALX1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ALX1. Recombinant protein control fragment (Product #RP-100682).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8092
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.