Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ALX3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA536138
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA536138 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA536138 Supplier Invitrogen™ Supplier No. PA536138
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 36 kDa. Purity is >95% by SDS-PAGE.

Aristaless-related genes are a group of paired-related homeobox genes which play a role in regulating vertebrate embryogenesis. The homeodomain transciption factor aristaless-like 3 (ALX3) is expressed in mouse embryos from 8 days of gestation, predominantly in neural crest-derived mesenchyme and in lateral plate mesoderm. Expression analysis of human and mouse tissue reveals predominant ALX3 expression in brain tissue. The Alx3 gene maps to chromosome 1p23-p13 and encodes a 343 amino acid protein. Preferential methylation of Alx3 occurs in advanced-stage neuroblastoma and may repress ALX3 expression. Treatment with the methylation inhibitor 5-aza-2'-deoxycytidine restores ALX3 expression. Alx3 (-) mice lack a phenotype distinct from wild-type mice, however Alx3/Alx4 double mutants demonstrate severe craniofacial abnormalities not present in Alx4 single mutants. Specifically, Alx3/Alx4 double mutant newborn mice have cleft nasal regions in addition to malformation of other neural crest-derived skull structures.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALX3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene ALX3
Gene Accession No. O70137, O95076
Gene Alias ALX homeobox 3; ALX3; aristaless 3; aristaless-like homeobox 3; FND; FND1; frontonasal dysplasia; homeobox protein aristaless-like 3; Proline-rich transcription factor ALX3
Gene Symbols ALX3
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 171-220 of human ALX3.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11694, 257, 365900
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.